Order Entry
Italy
ContactUsLinkComponent
Supelco® Organic Reference Standard, δ-BHC
Supelco® Organic Reference Standard, δ-BHC
Catalog # 40103-U
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, δ-BHC
Catalog # 40103-U
Supplier:  Merck
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    1
  • Standards:
    Certified reference material, 1000 mug/ml in methanol
  • CAS No.:
    319-86-8
  • Description:
    δ-BHC
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H225 - H301 + H311 + H331 - H370 - H412
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    δ-HCH solution; δ-1,2,3,4,5,6-Hexachlorocyclohexane solution
  • Formula:
    C₆H₆Cl₆
  • Molecular weight:
    290,83
  • Flash point:
    11 °C

Specifications

About this item

  • GHS signal: GHS02, GHS06, GHS08
Caution: P210 - P273 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311