L(+)-Tartaric acid 99.5-101.0%, powder Ph. Eur.
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:87-69-4
:
Formula:C₄H₆O₆
MDL Number:MFCD00064207
Merck Index:13,09158
Boiling Pt:399 °C (1013 hPa)
Melting Pt:168…170 °C
Density:1.759 g/cm³ (20 °C)
Flash Pt:150 °C (closed cup)
:150.09 g/mol
Storage Temperature:Ambient
EINECS:201-766-0
REACH:01-2119537204-47
:GHS05
Specification Test Results
- Assay (calculated on dried substance):99.5 - 101.0 %
- Appearance:Conforms (see CoA/CoS)
- Identification A:Passes test
- Identification B:Passes test
- Appearance of solution:Passes test
- Spec. opt. rotation (dried substance):12.0 - 12.8 °
- Oxalic acid:Max. 350 ppm
- Cl (Chloride):Max. 100 ppm
- SO4 (Sulphate):Max. 150 ppm
- Ca (Calcium):Max. 200 ppm
- Heavy metals (as Pb):Max. 10 ppm
- Loss on drying (105°C):Max. 0.2 %
- Sulphated ash:Max. 0.1 %
- Residual solvents:Unlikely by manuf.process
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 6L Plastic bottle for solids DIN 80 (ZPACK13256000)
Packaging Information
83511_L(+)-TARTARIC ACID POWDER PH.EUR_BSE_REV1.pdf
BSE/TSE Certificates
83511_L(+)-TARTARIC ACID POWDER PH.EUR_Nanomaterials_REV0.pdf L(+)-Tartaric acid 99,5 - 101%, Powder Ph. Eur.
Additional Product Details
spec_VWRC83511.290 spec_VWRC83511.290 spec_VWRC83511.360 spec_VWRC83511.360 spec_VWRC83511.460
Product specification
More
VWR Chemicals for Biopharma and Pharma Production
Product Literature
Frequently Bought Together