Organic reference standard, 2,2',3,3',4,5,5',6-Octachlorobiphenyl (PCB No. 198) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20019800IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:68194-17-2
- Description:2,2',3,3',4,5,5',6-Octachlorobiphenyl (PCB No. 198) 10 µg/ml in isooctane
- Formula:C₁₂H₂Cl₈
- Description:2,2',3,3',4,5,5',6-Octachlorobiphenyl (PCB No. 198)