Order Entry
Italy
ContactUsLinkComponent
 
Organic reference standard, 3-Chlorobiphenyl (PCB No. 2) 10 µg/ml in isooctane, Dr Ehrenstorfer
  EHERL20000200IO
 :  
Organic reference standard, 3-Chlorobiphenyl (PCB No. 2) 10 µg/ml in isooctane, Dr Ehrenstorfer
  EHERL20000200IO
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    2051-61-8
  • Description:
    3-Chlorobiphenyl (PCB No. 2) 10 µg/ml in isooctane
  • Formula:
    C₁₂H₉Cl
  • Description:
    3-Chlorobiphenyl (PCB No. 2)