Order Entry
Italy
ContactUsLinkComponent
 
Organic reference standard, 2,3',4,5',6-Pentachlorobiphenyl (PCB No. 121) 10 µg/ml in isooctane, Dr Ehrenstorfer
  EHERL20012100IO
 :  
Organic reference standard, 2,3',4,5',6-Pentachlorobiphenyl (PCB No. 121) 10 µg/ml in isooctane, Dr Ehrenstorfer
  EHERL20012100IO
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    56558-18-0
  • Description:
    2,3',4,5',6-Pentachlorobiphenyl (PCB No. 121) 10 µg/ml in isooctane
  • Formula:
    C₁₂H₅Cl₅
  • Description:
    2,3',4,5',6-Pentachlorobiphenyl (PCB No. 121)