Organic reference standard, 2,3,3',4',5,6-Hexachlorobiphenyl (PCB No. 163) 10 µg/ml in isooctane, Dr Ehrenstorfer
Codice catalogo EHERL20016300IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:74472-44-9
- Description:2,3,3',4',5,6-Hexachlorobiphenyl (PCB No. 163) 10 µg/ml in isooctane
- Formula:C₁₂H₄Cl₆
- Description:2,3,3',4',5,6-Hexachlorobiphenyl (PCB No. 163)