Order Entry
Italy
ContactUsLinkComponent
L(+)-Lithium lactate for synthesis, Sigma-Aldrich®
L(+)-Lithium lactate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
L(+)-Lithium lactate for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Molar Mass:
    96.01 g/mol

Specification Test Results

Related Information

Frequently Bought Together