Organic reference standard, 2,2',3,4,6'-Pentachlorobiphenyl (PCB No. 89) 10 µg/ml in isooctane, Dr Ehrenstorfer
EHERL20008900IO
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:73575-57-2
- Description:2,2',3,4,6'-Pentachlorobiphenyl (PCB No. 89) 10 µg/ml in isooctane
- Formula:C₁₂H₅Cl₅
- Description:2,2',3,4,6'-Pentachlorobiphenyl (PCB No. 89)