Organic reference standard, 2,3,3',4'-Tetrachlorobiphenyl (PCB No. 56) 10 µg/ml in isooctane, Dr Ehrenstorfer
EHERL20005600IO
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:41464-43-1
- Description:2,3,3',4'-Tetrachlorobiphenyl (PCB No. 56) 10 µg/ml in isooctane
- Formula:C₁₂H₆Cl₄
- Description:2,3,3',4'-Tetrachlorobiphenyl (PCB No. 56