
L(+)-Arginine
Catalog # PROOMM0568.00
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:174.2 g/mol
Merck Index:13,00785
Boiling Pt:368 °C (1013 hPa)
Melting Pt:238 °C
Density:1.3 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-811-1
:GHS07
CAS Number:74-79-3
Formula:C₆H₁₄N₄O₂
MDL Number:MFCD00002635