Organic reference standard, 2,2',3,4',5',6-Hexachlorobiphenyl (PCB No. 149) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20014900IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:38380-04-0
- Description:2,2',3,4',5',6-Hexachlorobiphenyl (PCB No. 149) 10 µg/ml in isooctane
- Formula:C₁₂H₄Cl₆
- Description:2,2',3,4',5',6-Hexachlorobiphenyl (PCB No. 149)