Order Entry
Italy
ContactUsLinkComponent
L(+)-Arginine 98+%
Irritant
L(+)-Arginine 98+%
Certificates
undefined
Irritant
L(+)-Arginine 98+%

Specification Test Results

  • Appearance:
    White to almost white Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.5 % (1 g, 105°C)
  • Residue after ignition:
    ≤0.3 %
  • Specific optical rotation:
    +25.8° to +28.5° (20°C, 589 nm) (c=8, 6 N HCl) (on dried substance)
  • Titration with HClO4:
    98.0 to 102.0 % (on dried substance)
  • Chloride (Cl):
    ≤500 ppm
  • Sulfate (SO4):
    ≤300 ppm
  • Iron (Fe):
    ≤30 ppm
  • Heavy metals (as Pb):
    ≤15 ppm

Specification Test Results

Related Information

Frequently Bought Together