Organic reference standard, 2,2',3,3',4,4',5,6'-Octachlorobiphenyl (PCB No. 196) 10 µg/ml in isooctane, Dr Ehrenstorfer
EHERL20019600IO
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:42740-50-1
- Description:2,2',3,3',4,4',5,6'-Octachlorobiphenyl (PCB No. 196) 10 µg/ml in isooctane
- Formula:C₁₂H₂Cl₈
- Description:2,2',3,3',4,4',5,6'-Octachlorobiphenyl (PCB No. 196)