Order Entry
Italy
ContactUsLinkComponent
Organic reference standard, 2,2',3,4,4',5,6-Heptachlorobiphenyl (PCB No. 181) 10 µg/ml in isooctane, Dr Ehrenstorfer
Organic reference standard, 2,2',3,4,4',5,6-Heptachlorobiphenyl (PCB No. 181) 10 µg/ml in isooctane, Dr Ehrenstorfer
Codice catalogo EHERL20018100IO
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, 2,2',3,4,4',5,6-Heptachlorobiphenyl (PCB No. 181) 10 µg/ml in isooctane, Dr Ehrenstorfer
Codice catalogo EHERL20018100IO
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    74472-47-2
  • Description:
    2,2',3,4,4',5,6-Heptachlorobiphenyl (PCB No. 181) 10 µg/ml in isooctane
  • Formula:
    C₁₂H₃Cl₇
  • Description:
    2,2',3,4,4',5,6-Heptachlorobiphenyl (PCB No. 181)

Specifications

About this item