N-Acetyl-L-cysteine, Millipore®
Catalog # 106425-5
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:163.2 g/mol
Merck Index:13,00090
Boiling Pt:371 °C (1013 hPa)
Melting Pt:106…108 °C
Density:1.48 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:210-498-3
CAS Number:616-91-1
Formula:C₅H₉NO₃S
MDL Number:MFCD00004880