Order Entry
Italy
ContactUsLinkComponent
N-Acetyl-L-cysteine 98%
N-Acetyl-L-cysteine 98%
Certificates
undefined
N-Acetyl-L-cysteine 98%
Certificates

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.5 %
  • Heavy metals:
    ≤10 ppm
  • Residue after ignition:
    ≤0.5 %
  • Specific optical rotation:
    +21° to +27° (20°C, 589 nm) (c=5, phosphate buffer pH 7) (on dried substance)
  • Titration with NaOH:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together