Order Entry
Italy
ContactUsLinkComponent
Pesticide reference standard, 1,2,3-Trichloropropane 200 µg/ml in methanol, Supelco®
Pesticide reference standard, 1,2,3-Trichloropropane 200 µg/ml in methanol, Supelco®
  48355.
 :  Merck
 :  
Pesticide reference standard, 1,2,3-Trichloropropane 200 µg/ml in methanol, Supelco®
  48355.
 :  Merck
 :  

 

  • Pk:
    1
  • CAS No.:
    96-18-4
  • Description:
    1,2,3-Trichloropropane 200 µg/ml in methanol
  • Formula:
    C₃H₅Cl₃
  • Description:
    1,2,3-Trichloropropane