
Platinium(IV) oxide hydrate 99.9% (71 - 75% Pt) (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:52785-06-5
:MFCD00011184
Formula:O₂Pt·H₂O
MDL Number:MFCD00066964
:245.09 g/mol
Merck Index:14,7527
Melting Pt:450 °C
Density:10.2 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-223-0
UN:1479
ADR:5.1,II
:GHS03
Frequently Bought Together