


Nickel(II) chloride hexahydrate, EMSURE® for analysis, Supelco®
Catalog # 1.06717.0250
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Solubility:553 g/l (20 °C)
- Melting Point:1001 °C (anhydrous substance),decomposes
- Molar Mass:237.66 g/mol
- Bulk Density:640 kg/m3
- Boiling Point:987 °C (sublimed),(anhydrous substance)
- pH:4.9 (100 g/l, H2O, 20 °C)
- Hazard Class:6.1
- Hazard Class Description:Toxic materials
Specification Test Results
About this item
:
:237.69 g/mol
Merck Index:13,06529
Melting Pt:140 °C
Density:1.84 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-743-0
UN:3288
ADR:6.1,III
:M-Clarity™ quality level = MQ300
:GHS06
:GHS08
:GHS09
CAS Number:7791-20-0
Formula:NiCl₂·6H₂O
MDL Number:MFCD00149809