Manganese(II) sulphate monohydrate 99.0-101.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
: Manganese sulphate monohydrate, Manganese(II) sulphate hydrate, Manganous sulphate monohydrate
:10034-96-5
:
:MnSO₄·1H₂O
:MFCD00149159
:13,05763
:700 °C
:2.95 g/cm³ (25 °C)
:232-089-9
:3077
:9,III
:169.02 g/mol
:GHS08
:GHS09
:Ambient
- Environmentally Preferable:
- Assay:99.0 - 101.0 %
- Insolubility in water:Max. 100 ppm
- Loss on ignition (500°C):10.0 - 12.0 %
- Substances reducing KMnO4:Passes test
- Cl (Chloride):Max. 10 ppm
- Ca (Calcium):Max. 50 ppm
- Heavy metals (as Pb):Max. 10 ppm
- Cu (Copper):Max. 5 ppm
- Fe (Iron):Max. 10 ppm
- K (Potassium):Max. 50 ppm
- Mg (Magnesium):Max. 50 ppm
- Na (Sodium):Max. 50 ppm
- Ni (Nickel):Max. 0.02 %
- Pb (Lead):Max. 10 ppm
- Zn (Zinc):Max. 50 ppm
- Conforms to ACS:Passes test
- Conforms to Reag. Ph.Eur.:Passes test
Frequently Bought Together