Order Entry
Italy
ContactUsLinkComponent
 
Copper(II) nitrate trihydrate, EMSURE® for analysis, Supelco®
 :  Merck
 
Copper(II) nitrate trihydrate, EMSURE® for analysis, Supelco®
 :  Merck
 

 

  • Solubility:
    2670 g/l (20 °C)
  • Melting Point:
    114 °C
  • Molar Mass:
    241.60 g/mol
  • Bulk Density:
    1050 kg/m3
  • Density:
    2.05 g/cm3 (20 °C)
  • pH:
    3 - 4 (50 g/l, H2O, 20 °C)
  • Hazard Class:
    5.1
  • Hazard Class Description:
    Oxidizer

 

Frequently Bought Together