Order Entry
Italy
ContactUsLinkComponent
Barium hydroxide octahydrate ≥98%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Irritant
Corrosive
Barium hydroxide octahydrate ≥98%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Corrosive
Barium hydroxide octahydrate ≥98%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (at filling):
    Min. 98 %
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Insolubility in diluted HCl:
    Max. 50 ppm
  • Not precipitated by dil. H2SO4 (as SO4):
    Max. 0.2 %
  • CO3 (as BaCO3) (at filling):
    Max. 2 %
  • Cl (Chloride):
    Max. 10 ppm
  • S (Sulphur):
    Max. 5 ppm
  • Ca (Calcium):
    Max. 30 ppm
  • Fe (Iron):
    Max. 5 ppm
  • Sr (Strontium):
    Max. 1.5 %
  • Conforms to Reag. Ph.Eur.:
    Passes test

Specification Test Results

Frequently Bought Together