Aluminium potassium sulphate dodecahydrate 98.0-102.0% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7784-24-9
:
Formula:AlK(SO₄)₂·12H₂O
MDL Number:MFCD00149143
:474.39 g/mol
Merck Index:14,360
Melting Pt:92 °C
Density:1.757
Storage Temperature:Ambient
EINECS:233-141-3
Related Information
Frequently Bought Together