Sodium dihydrogen phosphate monohydrate ≥98% ACS, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
Synonyms: Sodium phosphate monobasic monohydrate, Sodium dihydrogen orthophosphate monohydrate, Monosodium phosphate monohydrate, Sodium dihydrogenorthophosphate monohydrate, Sodium orthophosphate monobasic monohydrate
CAS Number:10049-21-5
:
Formula:NaH₂PO₄·H₂O
MDL Number:MFCD00149208
Molecular Weight:137.99 g/mol
Melting Pt:650 °C (anhydr.)
ADR:3
Merck Index:13,08734
Storage Temperature:Ambient
EINECS:231-449-2
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:98.0 - 102.0 %
- pH (5 %, 25 °C):4.1 - 4.5
- Water insoluble matter:Max. 0.01 %
- Calcium (Ca):Max. 50 ppm
- Iron (Fe):Max. 10 ppm
- Potassium (K):Max. 100 ppm
- Heavy metals (as Pb):Max. 0.001 %
- Chloride (Cl):Max. 5 ppm
- Sulfate (SO₄):Max. 30 ppm
Specification Test Results
Frequently Bought Together