
p-Toluenesulphonic acid monohydrate 99%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6192-52-5
:
Formula:C₇H₈O₃S·1H₂O
MDL Number:MFCD00142137
:190.22 g/mol
Merck Index:12,09671
Melting Pt:100…108 °C
Density:1,24 g/cm³ (20 °C)
Flash Pt:180 °C
Storage Temperature:Ambient
EINECS:203-180-0
UN:2585
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White to off-white Adhering crystals (May discolor to pink on storage)
- Infrared spectrum:Conforms
- Melting point:100 to 108 °C
- Titration Acid:The test method measures the total amount of different isomers of Toluenesulfonic acid (TSA).
- Titration with CHA:≥98.5 %
Specification Test Results
Related Information
Frequently Bought Together