Order Entry
Italy
ContactUsLinkComponent
EDTA disodium salt dihydrate, Molecular Biology Grade, Calbiochem®, Millipore®
EDTA disodium salt dihydrate, Molecular Biology Grade, Calbiochem®, Millipore®
Catalog # 324503-1
CAS Number:  
undefined
EDTA disodium salt dihydrate, Molecular Biology Grade, Calbiochem®, Millipore®
Catalog # 324503-1
CAS Number:  

Specifications

  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :372.24 g/mol
Merck Index:13,03543
Melting Pt:255 °C
Storage Temperature:Ambient
EINECS:205-358-3
CAS Number:6381-92-6
Formula:C₁₀H₁₄N₂Na₂O₈·2H₂O
MDL Number:MFCD00150037