tri-Sodium citrate dihydrate
Catalog # ICNA11TSC001KG
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:294.1 g/mol
Merck Index:13,08675
Melting Pt:150 °C
Density:1.76 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-675-3
CAS Number:6132-04-3
Formula:Na₃C₆H₅O₇·2H₂O
MDL Number:MFCD00150031