Order Entry
Italy
ContactUsLinkComponent
 
tri-Sodium citrate dihydrate
Catalog # MANA814029
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
tri-Sodium citrate dihydrate
Catalog # MANA814029
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Pk:
    2,5 g
  • Seq:
    3122

Specifications

About this item

 :
 :294.1 g/mol
Merck Index:13,08675
Melting Pt:150 °C
Density:1.76 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-675-3
CAS Number:6132-04-3
Formula:Na₃C₆H₅O₇·2H₂O
MDL Number:MFCD00150031