Order Entry
Italy
ContactUsLinkComponent
Magnesium acetate tetrahydrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Magnesium acetate tetrahydrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates
Magnesium acetate tetrahydrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    1200 g/l (15 °C)
  • Melting Point:
    80 °C
  • Molar Mass:
    214.46 g/mol
  • Bulk Density:
    510 kg/m3
  • Density:
    1.45 g/cm3
  • pH:
    6.1 (50 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Frequently Bought Together