Sodium acetate trihydrate 99.0-101.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:6131-90-4
:
Formula:H₃CCOONa·3H₂O
MDL Number:MFCD00071557
:136.08 g/mol
Merck Index:13,08642
Boiling Pt:120…123 °C (1013 hPa)
Melting Pt:58 °C
Density:1.4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-823-8
Specification Test Results
- Assay:99.0 - 101.0 %
- Appearance of solution:Passes test Ph.Eur.
- Identification A:Passes test
- Identification B:Passes test
- IR Spectrum:Passes test
- Solution in water:Passes test
- pH (20°C; 5 %):7.5 - 9.0
- pH (25°C; 5 %):7.5 - 9.0
- Heavy metals (as Pb):Max. 5 ppm
- Insolubility in water:Max. 50 ppm
- Loss on drying (130°C):39.0 - 40.5 %
- Substances reducing KMnO4 (as HCOOH):Max. 50 ppm
- Cl (Chloride):Max. 10 ppm
- PO4 (Phosphate):Max. 2 ppm
- SO4 (Sulphate):Max. 20 ppm
- Total N (Nitrogen):Max. 10 ppm
- Al (Aluminium):Max. 5 ppm
- As (Arsenic):Max. 2 ppm
- Ca (Calcium):Max. 10 ppm
- Ca + Mg (as Ca):Max. 50 ppm
- Cu (Copper):Max. 3 ppm
- Fe (Iron):Max. 5 ppm
- K (Potassium):Max. 50 ppm
- Mg (Magnesium):Max. 5 ppm
- Pb (Lead):Max. 5 ppm
- Conforms to ACS:Passes test
- Conforms to Reag. Ph.Eur.:Passes test
Specification Test Results
Frequently Bought Together