New fuchsine, Certistain® for microscopy, Sigma-Aldrich®
Catalog # 1.05226.0000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Solubility:10 g/l (25 °C)
- Molar Mass:365.91 g/mol
- Bulk Density:450 kg/m3
- pH:3 - 4 (10 g/l, H2O, 20 °C)
Specification Test Results
About this item
:
:365.91 g/mol
EINECS:221-831-7
:GHS08
Storage Temperature:Ambient
:1
CAS Number:3248-91-7
Formula:C₂₂H₂₄ClN₃
MDL Number:MFCD00012567