Buffer, TAE solution 10X concentrate (TRIS-acetate-EDTA buffer) pH 8,3, Millipore®
Catalog # 1.06023.1000
Supplier: Merck
CAS Number:
Jump to:
Product Details & Documents
Documents
ProductPage_106023 ProductPage_106023
Product specification
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Description:TAE buffer solution 10X concentrate (TRIS-acetate-EDTA buffer) pH 8,3
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



