Order Entry
Italy
ContactUsLinkComponent
 
Sodium hydroxide 0.1 mol/l (0.1 N) in ethanol, AVS TITRINORM
  83891.260DP
 :  
undefined
Sodium hydroxide 0.1 mol/l (0.1 N) in ethanol, AVS TITRINORM
  83891.260DP
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

 

 

  • Titer (20°C; real value 0.2 % accuracy):
    0.0998 - 0.1002 mol/l

 

 

 :
 :40 g/mol
 :Ambient
 :215-185-5
 :2924
 :3 (8),II
 :GHS02
 :1310-73-2
 :NaOH
 :MFCD00003548