Order Entry
Italy
ContactUsLinkComponent
Acrylamide 40% in aqueous solution, Millipore®
Health
Irritant
Acrylamide 40% in aqueous solution, Millipore®
Catalog # 1.00633.1000
Supplier:  Merck
CAS Number:  
undefined
Health
Irritant
Acrylamide 40% in aqueous solution, Millipore®
Catalog # 1.00633.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :71.08 g/mol
ADR:6.1,III
 :M-Clarity™ quality level = MQ200
EINECS:201-173-7
UN:3426
 :GHS08
 :GHS07
Storage Temperature:Refrigerator
CAS Number:79-06-1
Formula:C₃H₅NO
MDL Number:MFCD00008032