Order Entry
Italy
ContactUsLinkComponent
Mercaptoacetic acid (thioglycolic acid) ≥80% in aqueous solution, Supelco®
Toxic
Corrosive
Mercaptoacetic acid (thioglycolic acid) ≥80% in aqueous solution, Supelco®
Catalog # 88652-1L
Supplier:  Merck
CAS Number:  
undefined
Toxic
Corrosive
Mercaptoacetic acid (thioglycolic acid) ≥80% in aqueous solution, Supelco®
Catalog # 88652-1L
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :92.12 g/mol
Density:1.27 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1940
ADR:8,II
 :M-Clarity™ quality level = MQ100
EINECS:200-677-4
 :GHS06
 :GHS05
CAS Number:68-11-1
Formula:HSCH₂COOH
MDL Number:MFCD00004876