



Glutaraldehyde 25% in aqueous solution for electron microscopy, Sigma-Aldrich®
Catalog # 1.04239.0000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Assay (alkalimetric):23.5 - 26.5 %
- pH-value:≥ 3.0
- Density (d 20 °C/ 4 °C):1.060 - 1.065
- Absorbance A (235 nm; 1:200 deluted with water; 1 cm):≤ 0.3
- Density:1.06 g/cm3 (20 °C)
- pH:3.0 (H2O, 20 °C)
Specification Test Results
About this item
:
:100.12 g/mol
Boiling Pt:101 °C (1013 hPa)
Melting Pt:–6 °C
Density:1,06 g/cm³ (20 °C)
Flash Pt:≥ 71 °C
Storage Temperature:Refrigerator
EINECS:203-856-5
UN:1760
ADR:8,II
:M-Clarity™ quality level = MQ200
:GHS08
:GHS05
:GHS07
:GHS09
:1
CAS Number:111-30-8
Formula:C₅H₈O₂
MDL Number:MFCD00007025