Order Entry
Italy
ContactUsLinkComponent
(±)-3,7-Dimethyl-3-octanol 97+%
Irritant
(±)-3,7-Dimethyl-3-octanol 97+%
Certificates
undefined
Irritant
(±)-3,7-Dimethyl-3-octanol 97+%

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4310 to 1.4350 (20°C, 589 nm)
  • GC:
    ≥97.0 %

Specification Test Results

Related Information

Frequently Bought Together