

(±)-3,5,5-Trimethyl-1-hexanol ≥85.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:3452-97-9
:
Formula:(CH₃)₃CCH₂CH(CH₃)CH₂CH₂OH
MDL Number:MFCD00036138
Boiling Pt:193°C
Melting Pt:–70 °C
Density:0,83 g/cm³ (20 °C)
Flash Pt:79°C (closed cup)
Storage Temperature:Ambient
EINECS:222-376-7
ADR:9,III
:144.26 g/mol
UN:3082
:GHS07
:GHS08
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 85.0 %
Specification Test Results
Frequently Bought Together