(±)-1-Methoxy-2-propanol, EMPLURA®, Supelco®
: Merck
:107-98-2
:
:CH₃CH(OH)CH₂OCH₃
:MFCD00004537
:119.6 °C (1013 hPa)
:–97 °C
:0.92 g/cm³ (20 °C)
:31 °C
:203-539-1
:3092
:3,
:M-Clarity™ quality level = MQ200
:90.12 g/mol
:GHS02
:GHS07
:Ambient
Frequently Bought Together