2-Bromo-2-methylpropane stabilised for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: tert-Butyl bromide
CAS Number:507-19-7
:
Formula:C₄H₉Br
MDL Number:MFCD00000125
Boiling Pt:73
Density:1.2125 g/cm³ (20 °C)
:Stabilised with calcium carbonate
Molecular Weight:137.02 g/mol
Melting Pt:–20 °C
Flash Pt:16
Storage Temperature:Ambient
EINECS:208-065-9
UN:2342
ADR:3 II
Merck Index:13,01554
:M-Clarity™ quality level = MQ200
:GHS02
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:-20 °C
- Molar Mass:137.01 g/mol
- Boiling Point:72 - 74 °C
- Vapor Pressure:179.9 hPa (25 °C)
- Flash Point:16 °C
- Refractive Index:1.4279 (20 °C, 589 nm)
- Density:1.22 g/cm3 (20 °C)
Specification Test Results
Frequently Bought Together