Order Entry
Italy
ContactUsLinkComponent
Bromoform 99+% stabilised
Bromoform 99+% stabilised
Certificates
undefined
Bromoform 99+% stabilised
Specifications
  • Environmentally Preferable:
Specifications
Specification Test Results
  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • GC:
    ≥99.0 % (excluding stabilizer)
  • Stabilizer:
    ≤1.5 % w/w ethanol