Polysorbate 40
Supplier: Thermo Fisher Scientific
Jump to:
About this item
CAS No.: 9005-66-7 Molecular Formula: C62H122O26 Refractive Index: 1.4700 MDL No.: MFCD00165345
Synonyms: Sorbitan monopalmitate, ethoxylated, Polyoxyethylene sorbitan monopalmitate
- CAS Number:9005-66-7
- Formula:C₆₂H₁₂₂O₂₆
- MDL Number:MFCD00165345
- Density:1.090
- Storage Temperature:Ambient
2 Options Available Below
Product Details & Documents
CAS No.: 9005-66-7 Molecular Formula: C62H122O26 Refractive Index: 1.4700 MDL No.: MFCD00165345
Synonyms: Sorbitan monopalmitate, ethoxylated, Polyoxyethylene sorbitan monopalmitate
- CAS Number:9005-66-7
- Formula:C₆₂H₁₂₂O₂₆
- MDL Number:MFCD00165345
- Density:1.090
- Storage Temperature:Ambient



