Polyethylene glycol 400, Supelco®
81284-1G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
:
:400 g/mol
:∼250 °C (1013 hPa)
:4…8 °C
:1.13 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:245 °C
:Ambient
:500-038-2
:25322-68-3
:H(OCH₂CH₂)nOH
:MFCD00081839