Polyethylene glycol 400
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:25322-68-3
:
Formula:H(OCH₂CH₂)nOH
MDL Number:MFCD00081839
:400 g/mol
Merck Index:14,7568
Boiling Pt:∼250 °C (1013 hPa)
Melting Pt:4…8 °C
Density:1.126
Flash Pt:240 °C (464 °F)
Storage Temperature:Ambient
EINECS:500-038-2
Related Information
Frequently Bought Together