Platinum rhodium alloy (90:10 w%), wire for ISA type S standard grade thermocouple, Ø 0.25 mm (0.01 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
:
MDL Number:MFCD00198186
Storage Temperature:Ambient
Related Information
Frequently Bought Together