Order Entry
Italy
ContactUsLinkComponent
Platinum rhodium alloy (80:20 w%) ≥99.9% (metals basis), foil, Thickness: 0.1 mm (0.004 in)
Platinum rhodium alloy (80:20 w%) ≥99.9% (metals basis), foil, Thickness: 0.1 mm (0.004 in)
Catalog # 41623.FF
CAS Number:  
Platinum rhodium alloy (80:20 w%) ≥99.9% (metals basis), foil, Thickness: 0.1 mm (0.004 in)
Catalog # 41623.FF
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    1643
  • Pk Info:
    25×25 mm

Specifications

About this item

 :
Storage Temperature:Ambient
MDL Number:MFCD00198186