Platinum rhodium alloy (70:30 w%), wire for ISA type B standard grade thermocouple, Ø 0.5 mm (0.02 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
:
MDL Number:MFCD00198186
Storage Temperature:Ambient
Related Information
Frequently Bought Together