D-(+)-Galactose ≥98.5%, GPR RECTAPUR®
Catalog # 24333.296
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Assay:Min. 98.5 %
- Specific optical rotation (10 %; water):79 - 82 °
- Heavy metals (as Pb):Max. 10 ppm
- Ignition residue (SO4):Max. 0.1 %
- Loss on drying (100°C):Max. 1 %
Specification Test Results
About this item
:
:180.16 g/mol
Merck Index:13,04356
Boiling Pt:527 °C (1013 hPa)
Melting Pt:163…169 °C
Density:1.5 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-416-4
CAS Number:59-23-4
Formula:C₆H₁₂O₆
MDL Number:MFCD00151230