D-(+)-Sucrose, Millipore®
1.07651.9025
: Merck
:
- Pack type:Fibre carton
- Pk:25 kg
- Seq:4283
:
:13,08966
:169…170 °C
:1.29 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:342.3 g/mol
:Ambient
:200-334-9
:57-50-1
:C₁₂H₂₂O₁₁
:MFCD00006626