
D-Gluconic acid 50% in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:526-95-4
:MFCD00066366
Formula:C₆H₁₂O₇
MDL Number:MFCD00004240
:196.16 g/mol
Merck Index:14,4456
Density:1.234
Storage Temperature:Ambient
EINECS:208-401-4
UN:3265
ADR:8,III
:GHS05
Related Information
Frequently Bought Together